siRNA Details
Gene Symbol: Zgrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197997.2 | NP_932114.2 | 24 | 5356 | ||
GenBank mRNA | AK046429.1 | - | 19 | 3081 | ||
AK156938.1 | - | 9 | 1349 | |||
BC030185.1 | - | 4 | 410 | |||
BC079891.1 | - | 23 | 5468 | |||
BC082599.1 | - | 22 | 6394 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
