siRNA Details
Gene Symbol: Zgrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197997.2 | NP_932114.2 | 26 | 5997 | ||
GenBank mRNA | AK156938.1 | - | 11 | 1990 | ||
BC030185.1 | - | 6 | 1051 | |||
BC079891.1 | - | 25 | 6109 | |||
BC082599.1 | - | 24 | 7035 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
