siRNA Details
Gene Symbol: Coa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198006.4 | NP_932123.3 | 2 | 259 | ||
GenBank mRNA | AK018255.1 | - | 2 | 135 | ||
AK050432.1 | - | 2 | 165 | |||
AK166651.1 | - | 2 | 189 | |||
AK177444.1 | - | 2 | 80 | |||
AK218336.1 | - | 2 | 80 | |||
BB860009.1 | - | 2 | 210 | |||
BB863739.1 | - | 2 | 202 | |||
BC002137.1 | - | 2 | 166 | 3484305 | ||
BY779699.1 | - | 2 | 259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
