siRNA Details
Gene Symbol: Wdr70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081402.1 | NP_001074871.1 | 3 | 174 | ||
GenBank mRNA | AK159714.1 | - | 3 | 156 | ||
AK160933.1 | - | 3 | 109 | |||
BC037184.1 | - | 3 | 175 | |||
BC051482.1 | - | 3 | 141 | |||
BC061236.1 | - | 3 | 174 |
Gene Symbol: Gm1943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK170003.1 | - | 1 | 160 | ||
BC024407.1 | - | 1 | 153 |

More Information
Other Species Targeted

Gene Ontology Categories:
