siRNA Details
Gene Symbol: Gm1943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK170003.1 | - | 1 | 180 | ||
BC024407.1 | - | 1 | 173 |
Gene Symbol: Wdr70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081402.1 | NP_001074871.1 | 3 | 194 | ||
GenBank mRNA | AK014750.1 | - | 3 | 191 | ||
AK159714.1 | - | 3 | 176 | |||
AK160933.1 | - | 3 | 129 | |||
BC037184.1 | - | 3 | 195 | |||
BC051482.1 | - | 3 | 161 | |||
BC061236.1 | - | 3 | 194 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
