siRNA Details
Gene Symbol: Wdr70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014750.1 | - | 4 | 1089 | ||
AK167464.1 | - | 1 | 466 | |||
BC037184.1 | - | 4 | 1093 | |||
BC051482.1 | - | 4 | 1059 | |||
BC061236.1 | - | 4 | 1092 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
