siRNA Details
Gene Symbol: Ly6g6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023463.3 | NP_075952.1 | 3 | 312 | ||
GenBank mRNA | AJ315546.1 | - | 3 | 247 | ||
AJ315547.1 | - | 2 | 589 | |||
AK003375.1 | - | 3 | 313 | |||
BC116365.1 | - | 3 | 278 | 40100048 | ||
BC116366.1 | - | 3 | 276 | 40100049 |
