siRNA Details
Gene Symbol: Rsc1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023544.5 | NP_076033.4 | 1 | 1918 | ||
GenBank mRNA | AK006435.1 | - | 1 | 732 |
Gene Symbol: Ddi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017966.2 | NP_001017966.1 | 10 | 3303 | ||
GenBank mRNA | AK129015.1 | - | 3282 | |||
BC021415.1 | - | 10 | 3284 | |||
BC031491.1 | - | 3 | 2057 |

More Information
Other Species Targeted

Gene Ontology Categories:
