siRNA Details
Gene Symbol: 1810037I17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024461.2 | NP_077781.1 | 1, 2 | 270 | ||
GenBank mRNA | AK007721.1 | - | 1, 2 | 271 | ||
AK009748.1 | - | 1, 2 | 236 | |||
AK013091.1 | - | 1, 2 | 255 | |||
AK170776.1 | - | 1, 2 | 236 | |||
BC002135.1 | - | 1, 2 | 279 | 3484100 | ||
BC086913.1 | - | 1, 2 | 235 | 30121587 | ||
BC091739.1 | - | 1, 2 | 238 |
