siRNA Details
Gene Symbol: Ccdc53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122960.1 | NP_001116432.1 | 7 | 791 | ||
NM_026070.3 | NP_080346.1 | 7 | 794 | |||
GenBank mRNA | AK004206.1 | - | 7 | 709 | ||
AK009358.1 | - | 7 | 690 | |||
AK013835.1 | - | 7 | 799 | |||
AK158822.1 | - | 7 | 795 | |||
BC029256.1 | - | 7 | 691 | 3661472 |
