siRNA Details
Gene Symbol: ARID3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307939.1 | NP_001294868.1 | 2 | 521 | ||
NM_006465.3 | NP_006456.1 | 2 | 521 | |||
GenBank mRNA | AB208830.1 | - | 2 | 440 | ||
AF116846.1 | - | 2 | 453 | |||
AK298716.1 | - | 2 | 446 | |||
BC041792.1 | - | 2 | 449 | 5297774 | ||
BC045545.1 | - | 2 | 495 | |||
BC060824.1 | - | 2 | 434 |
