siRNA Details
Gene Symbol: 1700093K21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110133.1 | NP_001103603.1 | 3 | 702 | ||
NM_026105.3 | NP_080381.2 | 3 | 384 | |||
GenBank mRNA | AK007139.1 | - | 2 | 141 | ||
BC049567.1 | - | 3 | 384 | 6773879 |
