siRNA Details
Gene Symbol: Paxbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026110.2 | NP_080386.3 | 13, 14 | 2225 | ||
GenBank mRNA | AK007365.1 | - | 7, 8 | 930 | ||
AK011477.1 | - | 3, 4 | 309 | |||
AY033907.1 | - | 13, 14 | 2010 | |||
AY033908.1 | - | 13, 14 | 2027 | |||
BC014838.1 | - | 6, 7 | 710 | 3588322 | ||
BC027145.1 | - | 5, 6 | 645 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
