siRNA Details
Gene Symbol: 1110008F13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026124.3 | NP_080400.1 | 4 | 686 | ||
GenBank mRNA | AK002452.1 | - | 4 | 644 | ||
AK003568.1 | - | 4 | 681 | |||
AK078178.1 | - | 4 | 681 | |||
BC019445.1 | - | 4 | 624 | 5125733 |
