siRNA Details
Gene Symbol: Gm3404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243109.1 | NP_001230038.1 | 2 | 233 | ||
GenBank mRNA | AK015946.1 | - | 2 | 234 | ||
CJ047759.1 | - | 2 | 240 |
Gene Symbol: 4930449I24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026136.2 | NP_080412.1 | 234 | |||
GenBank mRNA | AK015434.1 | - | 235 | |||
BC147588.1 | - | 231 | 9056578 | |||
BC147604.1 | - | 231 | 9056594 |
Gene Symbol: Gm3402 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243111.1 | NP_001230040.1 | 233 |
Gene Symbol: Gm3409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243113.1 | NP_001230042.1 | 2 | 233 |
Gene Symbol: Gm3415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243114.1 | NP_001230043.1 | 256 |
Gene Symbol: Gm6370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243110.1 | NP_001230039.1 | 2 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
