siRNA Details
Gene Symbol: 3632451O06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026142.4 | NP_080418.2 | 2 | 357 | ||
GenBank mRNA | AK014402.1 | - | 2 | 320 | ||
AK014563.1 | - | 2 | 374 | |||
AK028178.1 | - | 2 | 345 | |||
AK028180.1 | - | 2 | 361 | |||
AK032093.1 | - | 2 | 358 | |||
AK049427.1 | - | 2 | 364 | |||
BC023359.1 | - | 2 | 342 | 5043946 |
