siRNA Details
Gene Symbol: Ikbip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026166.2 | NP_080442.2 | 3 | 598 | ||
GenBank mRNA | AK004670.1 | - | 3 | 556 | ||
CF171082.1 | - | 3 | 580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
