siRNA Details
Gene Symbol: Ergic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286560.1 | NP_001273489.1 | 13 | 2474 | ||
NM_026168.4 | NP_080444.1 | 14 | 2583 | |||
GenBank mRNA | AK004454.1 | - | 1 | 709 | ||
AK004661.1 | - | 14 | 2247 | |||
AK030045.1 | - | 14 | 2233 | |||
AK086727.1 | - | 14 | 2237 | |||
AK167391.1 | - | 14 | 2236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
