siRNA Details
Gene Symbol: Alg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026247.3 | NP_080523.2 | 4 | 1367 | ||
GenBank mRNA | AK014790.1 | - | 4 | 1312 | ||
AK086546.1 | - | 4 | 1260 | |||
AK089102.1 | - | 4 | 1331 |
