siRNA Details
Gene Symbol: 4930549C01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026300.2 | NP_080576.1 | 2 | 418 | ||
GenBank mRNA | AK015053.1 | - | 2 | 381 | ||
AK016074.1 | - | 2 | 419 | |||
BC048442.1 | - | 2 | 408 | 6704947 |
