siRNA Details
Gene Symbol: Dnajc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286972.1 | NP_001273901.1 | 5 | 669 | ||
GenBank mRNA | BC024335.1 | - | 5 | 577 | 4974988 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
