siRNA Details
Gene Symbol: Asprv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026414.2 | NP_080690.2 | 1 | 835 | ||
GenBank mRNA | AK004007.1 | - | 1 | 282 | ||
AK160494.1 | - | 1 | 776 | |||
BC057938.1 | - | 1 | 848 | |||
BC108357.1 | - | 1 | 733 | |||
BC119099.1 | - | 1 | 351 | |||
BC119101.1 | - | 1 | 351 | |||
DQ841260.1 | - | 1 | 835 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
