siRNA Details
Gene Symbol: 1810043G02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026431.2 | NP_080707.2 | 7 | 1035 | ||
GenBank mRNA | AK003097.1 | - | 7 | 1043 | ||
AK004032.1 | - | 7 | 1036 | |||
AK075794.1 | - | 7 | 1022 | |||
AK147511.1 | - | 2 | 5625 | |||
AK165293.1 | - | 4 | 3836 | |||
AK198996.1 | - | 1 | 256 | |||
BC006038.1 | - | 5 | 906 | |||
BC010330.1 | - | 7 | 1006 | 2651637 | ||
BC016605.1 | - | 6 | 1760 | |||
BC029982.1 | - | 7 | 1015 |
