siRNA Details
Gene Symbol: Zfp169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164575.1 | NP_001158047.1 | 3, 4 | 319 | ||
NM_001164576.1 | NP_001158048.1 | 3, 4 | 351 | |||
NM_026450.3 | NP_080726.3 | 3, 4 | 351 | |||
GenBank mRNA | AK086737.1 | - | 3, 4 | 352 | ||
AK162962.1 | - | 3, 4 | 251 | |||
BC060222.1 | - | 3, 4 | 309 | 30356624 | ||
CJ110773.1 | - | 3, 4 | 320 |
