siRNA Details
Gene Symbol: Zfp169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164575.1 | NP_001158047.1 | 5 | 3245 | ||
NM_001164576.1 | NP_001158048.1 | 6 | 1240 | |||
NM_026450.3 | NP_080726.3 | 5 | 3277 | |||
GenBank mRNA | AK086737.1 | - | 6 | 1241 | ||
BC060222.1 | - | 5 | 3235 | 30356624 | ||
DV046615.1 | - | 269 |
