siRNA Details
Gene Symbol: 1600012H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083880.1 | NP_001077349.1 | 2 | 318 | ||
NM_001083881.1 | NP_001077350.1 | 2 | 388 | |||
NM_001083882.1 | NP_001077351.1 | 1 | 570 | |||
NM_026451.2 | NP_080727.2 | 2 | 323 | |||
GenBank mRNA | AK005427.1 | - | 2 | 300 | ||
AK017276.1 | - | 1 | 500 | |||
AK018298.1 | - | 2 | 246 | |||
AK028052.1 | - | 2 | 324 | |||
AK076083.1 | - | 2 | 280 | |||
AK076441.1 | - | 527 | ||||
AK076476.1 | - | 2 | 243 | |||
AK083064.1 | - | 1 | 511 | |||
AK150772.1 | - | 2 | 343 | |||
AK167490.1 | - | 2 | 330 | |||
BB610149.1 | - | 2 | 324 | |||
BC027313.1 | - | 2 | 302 | 3710655 | ||
BC043303.1 | - | 1 | 531 | |||
BC043449.1 | - | 1 | 531 | |||
BC054359.1 | - | 2 | 320 | |||
BC071250.1 | - | 2 | 235 |

More Information
Other Species Targeted

Gene Ontology Categories:
