siRNA Details
Gene Symbol: Cspp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026493.3 | NP_080769.3 | 3, 4 | 341 | ||
GenBank mRNA | AK013065.1 | - | 3, 4 | 310 | ||
AK015133.1 | - | 3, 4 | 310 | |||
AK032310.1 | - | 3, 4 | 321 | |||
AK077173.1 | - | 3, 4 | 329 | |||
BC019581.1 | - | 3, 4 | 297 | |||
BC151042.1 | - | 3, 4 | 314 | 9087955 | ||
CN459376.1 | - | 3, 4 | 311 | |||
FJ008676.1 | - | 2, 3 | 168 | |||
FJ008677.1 | - | 2, 3 | 204 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
