siRNA Details
Gene Symbol: Cspp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026493.3 | NP_080769.3 | 7 | 786 | ||
GenBank mRNA | AK013065.1 | - | 7 | 755 | ||
AK015133.1 | - | 7 | 755 | |||
AK032310.1 | - | 8 | 790 | |||
AK077173.1 | - | 7 | 774 | |||
BC019581.1 | - | 7 | 742 | |||
BC151042.1 | - | 7 | 759 | 9087955 | ||
CN457021.1 | - | 1 | 54 | |||
FJ008676.1 | - | 7 | 637 | |||
FJ008677.1 | - | 6 | 649 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
