siRNA Details
Gene Symbol: Cspp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026493.3 | NP_080769.3 | 7 | 655 | ||
GenBank mRNA | AK013065.1 | - | 7 | 624 | ||
AK015133.1 | - | 7 | 624 | |||
AK032310.1 | - | 8 | 659 | |||
AK077173.1 | - | 7 | 643 | |||
BC019581.1 | - | 7 | 611 | |||
BC151042.1 | - | 7 | 628 | 9087955 | ||
CN459376.1 | - | 7 | 625 | |||
FJ008676.1 | - | 7 | 506 | |||
FJ008677.1 | - | 6 | 518 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
