siRNA Details
Gene Symbol: CCT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166285.1 | NP_001159757.1 | 3, 4 | 407 | ||
NM_006429.3 | NP_006420.1 | 2, 3 | 311 | |||
NR_029402.1 | 2, 3 | 311 | ||||
XM_011532478.2 | XP_011530780.1 | 3, 4 | 341 | |||
XM_011532479.1 | XP_011530781.1 | 2, 3 | 244 | |||
GenBank mRNA | AF026292.1 | - | 2, 3 | 236 | ||
AK098549.1 | - | 2, 3 | 229 | |||
AK222478.1 | - | 2, 3 | 219 | |||
AK291961.1 | - | 2, 3 | 257 | |||
AK293175.1 | - | 2, 3 | 229 | |||
AK297846.1 | - | 3, 4 | 335 | |||
BC019296.1 | - | 2, 3 | 224 | 2905446 | ||
BC088351.1 | - | 2, 3 | 266 | 6202511 | ||
BM918058.1 | - | 287 | ||||
CR456719.1 | - | 1, 2 | 168 | |||
CR536511.1 | - | 1, 2 | 168 | |||
DC362157.1 | - | 2, 3 | 311 | |||
DC395264.1 | - | 2, 3 | 309 | |||
U83843.1 | - | 1, 2 | 132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
