siRNA Details
Gene Symbol: Wls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026582.4 | NP_080858.3 | 3 | 727 | ||
GenBank mRNA | AK019886.1 | - | 4 | 766 | ||
AK029053.1 | - | 3 | 686 | |||
AK029104.1 | - | 3 | 684 | |||
AK029315.1 | - | 3 | 684 | |||
AK031444.1 | - | 3 | 684 | |||
AK044810.1 | - | 3 | 686 | |||
AK049842.1 | - | 3 | 684 | |||
AK077407.1 | - | 3 | 685 | |||
AK128912.1 | - | 3 | 683 | |||
AK159207.1 | - | 3 | 684 | |||
AK159977.1 | - | 3 | 686 | |||
BC018381.1 | - | 2 | 311 | 4237619 | ||
BC075615.1 | - | 3 | 727 | 5703610 |
