siRNA Details
Gene Symbol: Wls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026582.4 | NP_080858.3 | 4 | 768 | ||
GenBank mRNA | AK019886.1 | - | 5 | 807 | ||
AK029053.1 | - | 4 | 727 | |||
AK029104.1 | - | 4 | 725 | |||
AK029315.1 | - | 4 | 725 | |||
AK031444.1 | - | 4 | 725 | |||
AK044810.1 | - | 4 | 727 | |||
AK049842.1 | - | 4 | 725 | |||
AK077407.1 | - | 4 | 726 | |||
AK128912.1 | - | 4 | 724 | |||
AK159207.1 | - | 4 | 725 | |||
AK159977.1 | - | 4 | 727 | |||
AK198815.1 | - | 2 | 50 | |||
BC018381.1 | - | 3 | 352 | 4237619 | ||
BC075615.1 | - | 4 | 768 | 5703610 |
