siRNA Details
Gene Symbol: Tmco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039483.1 | NP_001034572.1 | 7 | 1243 | ||
GenBank mRNA | AK004504.1 | - | 7 | 1085 | ||
BC011457.1 | - | 7 | 1117 | 3585288 | ||
BC019761.1 | - | 7 | 1161 | 5009960 | ||
BC020098.1 | - | 7 | 1173 | 4019484 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
