siRNA Details
Gene Symbol: REG1CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002714.1 | 1 | 1607 | |||
GenBank mRNA | AK126561.1 | - | 1 | 1607 | ||
D56494.1 | - | 4 | 362 | |||
D56495.1 | - | 4 | 367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
