siRNA Details
Gene Symbol: Zfp397 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027007.2 | NP_081283.2 | 2 | 644 | ||
GenBank mRNA | AK013086.1 | - | 2 | 607 | ||
AK032001.1 | - | 2 | 647 | |||
AK078454.1 | - | 1 | 2119 | |||
AY338468.1 | - | 1 | 468 | |||
BC021456.1 | - | 2 | 628 |
