siRNA Details
Gene Symbol: Lnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110209.1 | NP_001103679.1 | 14 | 1401 | ||
NM_027133.3 | NP_081409.1 | 13 | 1318 | |||
GenBank mRNA | AK014897.1 | - | 14 | 1298 | ||
AK048501.1 | - | 4 | 1561 | |||
AK136506.1 | - | 13 | 1293 | |||
AK137445.1 | - | 14 | 1401 | |||
AK173241.1 | - | 13 | 1250 | |||
AY312281.1 | - | 13 | 1239 | |||
BC057961.1 | - | 13 | 1245 | 5353629 | ||
BC060153.1 | - | 12 | 1157 |
