siRNA Details
Gene Symbol: Mettl7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027334.3 | NP_081610.2 | 2 | 1237 | ||
GenBank mRNA | AB253943.1 | - | 2 | 1205 | ||
AJ536232.1 | - | 2 | 1209 | |||
AK076169.1 | - | 2 | 1239 | |||
BC033446.1 | - | 2 | 1214 | |||
BC047149.1 | - | 2 | 1222 | |||
BC055034.1 | - | 2 | 1228 | 6306873 | ||
BC060144.1 | - | 2 | 1217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
