siRNA Details
Gene Symbol: Mettl7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027334.3 | NP_081610.2 | 2 | 1366 | ||
GenBank mRNA | AB253943.1 | - | 2 | 1334 | ||
AJ536232.1 | - | 2 | 1338 | |||
AK076169.1 | - | 2 | 1368 | |||
BC033446.1 | - | 2 | 1343 | |||
BC047149.1 | - | 2 | 1351 | |||
BC055034.1 | - | 2 | 1357 | 6306873 | ||
BC060144.1 | - | 2 | 1346 |
Gene Symbol: Mettl7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199477.2 | NP_955771.2 | 2 | 1343 | ||
GenBank mRNA | AB253944.1 | - | 2 | 1334 | ||
AK143923.1 | - | 2 | 1344 | |||
BC090971.1 | - | 2 | 1343 | 30314995 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
