siRNA Details
Gene Symbol: Tmem33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285452.1 | NP_001272381.1 | 7 | 1162 | ||
NM_028975.4 | NP_083251.2 | 8 | 1005 | |||
GenBank mRNA | AK005493.1 | - | 8 | 897 | ||
AK007607.1 | - | 8 | 813 | |||
AK007750.1 | - | 8 | 894 | |||
AK009040.1 | - | 3 | 237 | |||
AK032555.1 | - | 8 | 912 | |||
AK153441.1 | - | 8 | 830 | |||
AK165369.1 | - | 8 | 807 | |||
BC005562.1 | - | 8 | 806 | 3965864 | ||
BC016570.1 | - | 7 | 878 | 3583937 | ||
BC023966.1 | - | 7 | 871 | 5342147 |

More Information
Other Species Targeted

Gene Ontology Categories:
