siRNA Details
Gene Symbol: Zfp639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161818.1 | NP_001155290.1 | 7 | 700 | ||
NM_144519.4 | NP_653102.1 | 7 | 801 | |||
GenBank mRNA | AK049934.1 | - | 7 | 680 | ||
AK076145.1 | - | 7 | 802 | |||
AK076837.1 | - | 7 | 677 | |||
AK078000.1 | - | 7 | 701 | |||
AY736186.1 | - | 4 | 386 | |||
BC003941.1 | - | 5 | 450 | 3487151 | ||
BC042660.1 | - | 1 | 42 | |||
DQ323614.1 | - | 3 | 240 |
