siRNA Details
Gene Symbol: PDLIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011513.3 | NP_001011513.3 | 13 | 2030 | ||
NM_001256425.1 | NP_001243354.1 | 12 | 1938 | |||
NM_001256426.1 | NP_001243355.1 | 17 | 2444 | |||
NM_001256428.1 | NP_001243357.1 | 12 | 2205 | |||
NM_006457.4 | NP_006448.4 | 13 | 2357 | |||
XM_005262693.4 | XP_005262750.1 | 17 | 2741 | |||
XM_005262695.4 | XP_005262752.1 | 16 | 2723 | |||
XM_005262696.4 | XP_005262753.1 | 16 | 2341 | |||
XM_005262698.4 | XP_005262755.1 | 14 | 1970 | |||
XM_006714066.3 | XP_006714129.1 | 18 | 2756 | |||
XM_006714068.3 | XP_006714131.1 | 18 | 2374 | |||
XM_006714069.3 | XP_006714132.1 | 15 | 2360 | |||
XM_006714070.3 | XP_006714133.1 | 18 | 2357 | |||
XM_011531543.2 | XP_011529845.1 | 17 | 2339 | |||
XM_017007657.1 | XP_016863146.1 | 14 | 2345 | |||
XM_017007658.1 | XP_016863147.1 | 15 | 1978 | |||
GenBank mRNA | AB209531.1 | - | 9 | 2069 | ||
AF061258.1 | - | 13 | 2259 | |||
AL832157.1 | - | 13 | 1932 | |||
AL832686.1 | - | 17 | 2348 | |||
BC008741.2 | - | 13 | 2296 | 3345715 | ||
BC026029.2 | - | 11 | 1800 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
