siRNA Details
Gene Symbol: Gm40812 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK157581.1 | - | 1 | 1438 | ||
BC019819.1 | - | 1 | 943 | |||
BC049268.1 | - | 1 | 1279 | |||
BC056646.1 | - | 1 | 1412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
