siRNA Details
Gene Symbol: Gm42035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133569.1 | - | 465 | |||
FJ654104.1 | - | 1777 | ||||
FJ654105.1 | - | 1777 | ||||
FJ654106.1 | - | 1777 | ||||
FJ654107.1 | - | 1777 | ||||
FJ654108.1 | - | 1777 | ||||
FJ654112.1 | - | 1777 | ||||
FJ654131.1 | - | 1776 | ||||
FJ654132.1 | - | 1777 |
Gene Symbol: Gm3579 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036817.1 | - | 1 | 1921 | ||
AK037011.1 | - | 1 | 1921 | |||
AK158167.1 | - | 1 | 1919 | |||
AY140896.1 | - | 1 | 1776 | |||
BG084186.1 | - | 1 | 389 |
Gene Symbol: Gm40812 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK157581.1 | - | 1 | 2017 | ||
BC019819.1 | - | 1 | 1522 | |||
BC049268.1 | - | 1 | 1858 | |||
BC056646.1 | - | 1 | 1991 |
Gene Symbol: LOC67527 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004927.1 | - | 1920 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
