siRNA Details
Gene Symbol: Rnls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146342.2 | NP_001139814.2 | 3, 4 | 672 | ||
NM_001167818.1 | NP_001161290.1 | 5, 6 | 921 | |||
GenBank mRNA | AK014866.1 | - | 5, 6 | 790 | ||
AK018322.1 | - | 5, 6 | 889 | |||
AK170321.1 | - | 3, 4 | 673 | |||
BC052355.1 | - | 5, 6 | 752 | 6770333 | ||
DQ788834.1 | - | 5, 6 | 698 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
