siRNA Details
Gene Symbol: Rnls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146342.2 | NP_001139814.2 | 2 | 390 | ||
NM_001167818.1 | NP_001161290.1 | 4 | 639 | |||
GenBank mRNA | AK014866.1 | - | 4 | 508 | ||
AK018322.1 | - | 4 | 610 | |||
BC052355.1 | - | 4 | 470 | 6770333 | ||
DQ788834.1 | - | 4 | 416 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
