siRNA Details
Gene Symbol: Mypn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182992.2 | NP_892037.2 | 18 | 3861 | ||
GenBank mRNA | AK039718.1 | - | 2 | 932 | ||
AK220521.1 | - | 18 | 3861 | |||
BC052872.1 | - | 4 | 529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
