siRNA Details
Gene Symbol: 1700020N01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006179.1 | - | 2, 3 | 327 | ||
AK007146.1 | - | 2, 3 | 326 | |||
BC099546.1 | - | 2, 3 | 332 | 30920524 | ||
CA464995.1 | - | 2, 3 | 341 |
