siRNA Details
Gene Symbol: Osbpl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252489.1 | NP_001239418.1 | 15 | 2190 | ||
NM_001252490.1 | NP_001239419.1 | 14 | 2064 | |||
NM_001252491.1 | NP_001239420.1 | 13 | 2270 | |||
NM_001252492.1 | NP_001239421.1 | 14 | 2433 | |||
NM_001252493.1 | NP_001239422.1 | 15 | 2341 | |||
NM_207530.3 | NP_997413.2 | 28 | 3319 | |||
GenBank mRNA | AB017026.1 | - | 12 | 2314 | ||
AK133806.1 | - | 12 | 1696 | |||
AK168366.1 | - | 14 | 2394 | |||
BC076637.1 | - | 14 | 2064 | |||
BC092247.1 | - | 13 | 2270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
