siRNA Details
Gene Symbol: Ttyh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018148.1 | - | 14 | 2103 | ||
AK031550.1 | - | 12 | 2053 | |||
AK042758.1 | - | 14 | 1892 | |||
AK043998.1 | - | 14 | 2159 | |||
AK048413.1 | - | 14 | 1625 | |||
AK049903.1 | - | 13 | 1845 | |||
AK081981.1 | - | 14 | 2113 | |||
AK083126.1 | - | 13 | 1751 | |||
BC046310.1 | - | 14 | 1612 | 6307420 | ||
BC065694.1 | - | 13 | 2057 | 6813365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
