siRNA Details
Gene Symbol: Shank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ245904.1 | - | 15, 16 | 1962 | ||
HQ405763.1 | - | 1, 2 | 498 | |||
KF612593.1 | - | 1, 2 | 266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
